Assay Details
| Gene Symbol: Bhmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016668.3 | NP_057877.1 | 1 - 2 | 290 | 139 | |
| GenBank mRNA | AK145668.1 | - | 1 - 2 | 139 | ||
| BC110307.1 | - | 1 - 2 | 493 | 5310035 | 139 | |
| BY773713.1 | - | 1 - 2 | 291 | 139 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK145668.1, BC110307.1, NM_016668.3 transcripts.
Set Membership: |
|

Bhmt
Genomic Map