Assay Details
| Gene Symbol: Pin4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027181.1 | NP_081457.1 | 3 - 4 | 296 | 83 | |
| GenBank mRNA | AK010338.1 | - | 3 - 4 | 265 | 83 | |
| BC119355.1 | - | 3 - 4 | 296 | 8734104 | 83 | |
| BC119357.1 | - | 3 - 4 | 296 | 8734106 | 83 | |
More Information
Set Membership: |
|

Pin4
Genomic Map