Assay Details
| Gene Symbol: Ap3m1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018829.4 | NP_061299.3 | 1 - 2 | 302 | 163 | |
| GenBank mRNA | AF242857.1 | - | 1 - 2 | 288 | 163 | |
| BY774570.1 | - | 1 - 2 | 310 | 170 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF242857.1, NM_018829.4 transcripts.
Set Membership: |
|

Ap3m1
Genomic Map