Assay Details
| Gene Symbol: Psg25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_054060.1 | NP_473401.1 | 4 - 5 | 1322 | 95 | |
| GenBank mRNA | BC100352.1 | - | 4 - 5 | 1322 | 30715445 | 95 |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2067767 | BC100352.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2067767 | NM_054060.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 35361 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 227683 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 196017 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 133391 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 119065 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 539063 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P141121 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2067767 | BC100352.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2067767 | NM_054060.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24989003 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24989018 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24989024 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24989047 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24989012 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24989016 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24989030 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24989005 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24989033 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24989023 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24989032 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24989038 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24989009 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24989039 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24989008 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24989007 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24989049 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24989006 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24989055 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24988915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24989039 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24988956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24988979 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24988960 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24989055 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24989049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24989047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24989009 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24988990 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24988951 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24989024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24988953 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24989023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24989003 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24988949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24988946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24989016 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24989021 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24988921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24988977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24988980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24988986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24988926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24989005 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24988961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24988965 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24988920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24989033 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24988992 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24989018 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24988935 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24988972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24989012 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24989032 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24989038 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24989007 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24988901 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24988918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24988983 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24988941 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24989030 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24989006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24988981 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24988923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24988957 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24988924 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24988975 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24988991 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24989016 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06281260 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24988992 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06281242 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06530835 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06196483 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24989038 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24989018 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24989033 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24989055 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06530851 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24988977 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24989030 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24989000 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06281235 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06563722 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24989024 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24989023 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24989032 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24989039 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24988979 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24989059 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24989054 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06115903 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24989049 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24989021 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24988986 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06590437 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24989012 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24989060 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06530848 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24988980 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24988972 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24988975 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24988993 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24989005 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06256737 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06530841 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24988990 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24989007 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24989003 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06668158 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24988982 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10560376 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10560377 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10560379 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10560378 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10560380 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10560375 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700021603.mm.1 | NM_054060.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700021603.mm.1 | NM_054060.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700021603.mm.1 | TC0700002454.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700021602.mm.1 | NM_054060.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700021602.mm.1 | NM_054060.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700021602.mm.1 | TC0700002454.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 579119 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseRef-8 v2 Expression BeadChip | ILMN_2996933 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2996933 | - | - |

Psg25
Genomic Map