Assay Details
| Gene Symbol: Selt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040396.2 | NP_001035486.2 | 3 - 4 | 454 | 82 | |
| GenBank mRNA | AK013022.1 | - | 3 - 4 | 418 | 82 | |
| AK032838.1 | - | 3 - 4 | 431 | 82 | ||
| AK047958.1 | - | 3 - 4 | 432 | 82 | ||
| AK146778.1 | - | 3 - 4 | 342 | 82 | ||
| BC019970.1 | - | 3 - 4 | 402 | 4458171 | 82 | |
| BC038867.1 | - | 3 - 4 | 330 | 1547418 | 82 | |
| BC043068.1 | - | 340 | 82 | |||
| BC132453.1 | - | 3 - 4 | 420 | 40130730 | 82 | |
| BC138424.1 | - | 3 - 4 | 420 | 8861444 | 82 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Selt
Genomic Map