Assay Details
| Gene Symbol: Selt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040396.2 | NP_001035486.2 | 4 - 5 | 543 | 79 | |
| GenBank mRNA | AK013022.1 | - | 4 - 5 | 507 | 79 | |
| AK032838.1 | - | 4 - 5 | 520 | 79 | ||
| AK047958.1 | - | 4 - 5 | 521 | 79 | ||
| AK146778.1 | - | 4 - 5 | 431 | 79 | ||
| BC019970.1 | - | 4 - 5 | 491 | 4458171 | 79 | |
| BC038867.1 | - | 4 - 5 | 419 | 1547418 | 79 | |
| BC043068.1 | - | 429 | 79 | |||
| BC132453.1 | - | 4 - 5 | 509 | 40130730 | 79 | |
| BC138424.1 | - | 4 - 5 | 509 | 8861444 | 79 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Selt
Genomic Map