Assay Details
| Gene Symbol: Ap2m1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK159534.1 | - | 4 - 5 | 567 | 148 | |
| AK172889.1 | - | 4 - 5 | 520 | 148 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ap2m1
Genomic Map