| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0700004004.mm.1 |
NM_013647.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700004004.mm.1 |
NM_013647.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700004004.mm.1 |
TC0700000495.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700004003.mm.1 |
NM_013647.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700004003.mm.1 |
NM_013647.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700004003.mm.1 |
TC0700000495.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700003995.mm.1 |
NM_013647.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700003995.mm.1 |
NM_013647.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700003995.mm.1 |
TC0700000495.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700003998.mm.1 |
NM_013647.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700003998.mm.1 |
NM_013647.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700003998.mm.1 |
TC0700000495.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700004000.mm.1 |
NM_013647.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700004000.mm.1 |
NM_013647.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700004000.mm.1 |
TC0700000495.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700004001.mm.1 |
NM_013647.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700004001.mm.1 |
NM_013647.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700004001.mm.1 |
TC0700000495.mm.1 |
MTA TC |