Assay Details
| Gene Symbol: Eno1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023119.2 | NP_075608.2 | 1 - 2 | 143 | 166 | |
| GenBank mRNA | BU526382.1 | - | 1 - 2 | 146 | 166 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_023119.2 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23404995 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23404989 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23404994 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23404993 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23404992 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23404997 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23404990 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23404991 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1958097 | BC004017.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1958097 | BC083334.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1958097 | BC085098.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1958097 | BC039179.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1958097 | BC056611.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 774251 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 67612 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 114310 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 191078 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 471648 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 384789 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 551203 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 495243 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 12163 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P166796 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P167214 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1958097 | BC004017.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1958097 | BC083334.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1958097 | BC085098.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1958097 | BC039179.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1958097 | BC056611.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P349939 | NM_001025388.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P349939 | BC004017.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P349939 | BC089539.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P349939 | BC083334.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P349939 | BC085098.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23405005 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23404984 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23405000 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23404964 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23404985 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23404993 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23404966 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23405013 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23404987 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23404962 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23404976 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23404998 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23405008 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23405006 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23405012 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23404970 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23405003 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23405001 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23405010 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23404975 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23404990 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23404997 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23404995 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23404980 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23404982 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23404991 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23404978 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23404988 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23404972 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P166796 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P167214 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05449258 | - | - |
| Whole Mouse Genome G4122A | A_52_P349939 | NM_001025388.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P349939 | BC004017.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P349939 | BC089539.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P349939 | BC083334.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P349939 | BC085098.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P106342 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P131709 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P114762 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P166796 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P167214 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03214004 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03213988 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03213990 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03213991 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03213994 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23404992 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23404990 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23404995 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23404993 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23404997 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23404991 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23404989 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23404994 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404973 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404959 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404947 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404950 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404962 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404951 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404976 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404975 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404988 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404982 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404990 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404991 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23404955 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23404975 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23404945 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23404962 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23404982 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03745296 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03954966 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23404966 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23404953 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23404980 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23404954 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23404959 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23404969 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23404941 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23404978 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23404993 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23404972 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23404984 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23404955 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23404950 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23404958 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23404948 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23404964 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03942718 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23404985 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23404986 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03823250 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23404976 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04005495 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23404947 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23404988 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03213988 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03534743 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23404973 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10510548 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10510550 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10510551 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10455597 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10455598 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10510549 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10473241 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1419023_PM_x_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10473240 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10510546 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10455595 | - | - |
| Mouse Genome 430 2.0 Array | 1419023_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1419023_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1419022_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1419022_PM_a_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0400016159.mm.1 | NM_023119.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400016159.mm.1 | NM_023119.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400016159.mm.1 | TC0400001974.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400016172.mm.1 | NM_023119.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400016172.mm.1 | NM_023119.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400016172.mm.1 | TC0400001974.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400016176.mm.1 | NM_023119.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400016176.mm.1 | NM_023119.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400016176.mm.1 | TC0400001974.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400016187.mm.1 | NM_023119.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400016187.mm.1 | NM_023119.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400016187.mm.1 | TC0400001974.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400016155.mm.1 | NM_023119.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400016155.mm.1 | NM_023119.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400016155.mm.1 | TC0400001974.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400016171.mm.1 | NM_023119.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400016171.mm.1 | NM_023119.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400016171.mm.1 | TC0400001974.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400016173.mm.1 | NM_023119.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400016173.mm.1 | NM_023119.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400016173.mm.1 | TC0400001974.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400016183.mm.1 | NM_023119.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400016183.mm.1 | NM_023119.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400016183.mm.1 | TC0400001974.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400016184.mm.1 | NM_023119.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400016184.mm.1 | NM_023119.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400016184.mm.1 | TC0400001974.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400016161.mm.1 | NM_023119.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400016161.mm.1 | NM_023119.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400016161.mm.1 | TC0400001974.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400016186.mm.1 | NM_023119.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400016186.mm.1 | NM_023119.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400016186.mm.1 | TC0400001974.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400016166.mm.1 | NM_023119.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400016166.mm.1 | NM_023119.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400016166.mm.1 | TC0400001974.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400016169.mm.1 | NM_023119.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400016169.mm.1 | NM_023119.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400016169.mm.1 | TC0400001974.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400016179.mm.1 | NM_023119.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400016179.mm.1 | NM_023119.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400016179.mm.1 | TC0400001974.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400016148.mm.1 | NM_023119.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400016148.mm.1 | NM_023119.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400016148.mm.1 | TC0400001974.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400016181.mm.1 | NM_023119.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400016181.mm.1 | NM_023119.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400016181.mm.1 | TC0400001974.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1427404_PM_x_at | - | - |
| Mouse Expression Set 430 | 1432829_PM_at | - | - |
| Mouse Expression Set 430 | 1419023_x_at | - | - |
| Mouse Expression Set 430 | 1427404_x_at | - | - |
| Mouse Expression Set 430 | 1419022_a_at | - | - |
| Mouse Expression Set 430 | 1432829_at | - | - |
| Mouse Expression Set 430 | 1432828_at | - | - |
| Mouse Expression Set 430 | 1432828_PM_at | - | - |
| Mouse Expression Set 430 | 1419023_PM_x_at | - | - |
| Mouse Expression Set 430 | 1419022_PM_a_at | - | - |
| Mouse Expression Set 430 | 1431111_at | - | - |
| Mouse Expression Set 430 | 1431111_PM_at | - | - |
| Murine Genome U74v2 Set | 160568_PM_at | - | - |
| Murine Genome U74v2 Set | 160568_at | - | - |
| Murine Genome U74v2 Set | 169423_i_at | - | - |
| Murine Genome U74v2 Set | 169423_PM_i_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427404_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1432829_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419023_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1432829_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427404_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419022_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419022_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1432828_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419023_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1432828_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419023_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419023_PM_x_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5340128 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 106290373 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5340128 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005340128 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1215995 | - | - |
| mouse-6 v1.1 (Illumina) | scl013806.12_49-S | - | - |
| mouse-6 v1.1 (Illumina) | scl4379.1.1_298-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 106290373 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5340128 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl013806.12_49-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 106290373 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5340128 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2958159 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1215995 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2628271 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2958159 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl013806.12_49-S | - | - |

Eno1
Genomic Map