Assay Details
| Gene Symbol: Oser1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025699.2 | NP_079975.2 | 4 | 1423 | 97 | |
| GenBank mRNA | AK088622.1 | - | 4 | 1442 | 97 | |
| AK139692.1 | - | 1595 | 97 | |||
| AK153499.1 | - | 4 | 1460 | 97 | ||
| BC005579.1 | - | 4 | 1423 | 3968876 | 97 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21812326 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21812330 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21812331 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21812334 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21812329 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21812328 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21812335 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2001159 | BC005579.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2001159 | AK088622.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2001159 | AK153499.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2001159 | NM_025699.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 515056 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 669697 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 573287 | AB032591.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 573287 | AK039764.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 573287 | AK044552.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 573287 | AK083141.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 573287 | NM_021483.2 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 222412 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 863924 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 296712 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 534572 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 564604 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 958508 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 403955 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 499875 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 873574 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 3864 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 147337 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2001159 | BC005579.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2001159 | AK088622.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2001159 | AK153499.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2001159 | NM_025699.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21812361 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21812324 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21812356 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21812346 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21812342 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21812349 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21812353 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21812343 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21812359 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21812332 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21812357 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21812354 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21812327 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21812355 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21812338 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21812331 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21812329 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21812313 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21812337 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21812322 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21812316 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21812340 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21812345 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21812315 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21812351 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21812310 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21812318 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21812334 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21812312 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04769505 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04769520 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04769535 | - | - |
| Whole Mouse Genome G4122A | A_51_P451808 | BC005579.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P451808 | AK014323.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P451808 | AK088622.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P451808 | AK139692.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P451808 | AK153499.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P129849 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P118720 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P04769505 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21812326 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21812334 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21812331 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21812335 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21812328 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21812329 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21812330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812179 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812322 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812275 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812199 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812276 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812316 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812380 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812231 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812236 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812342 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812329 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812195 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812278 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812332 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812283 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812280 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812205 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812359 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812338 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812365 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812367 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812293 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812331 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812193 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812237 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812255 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812357 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812339 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812356 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812257 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812180 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812233 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812302 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812260 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812186 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812318 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812269 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812196 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812281 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812298 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812287 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812373 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812312 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812203 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812239 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812362 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812296 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812364 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812344 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812235 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812274 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812303 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812291 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812361 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812267 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812189 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812194 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812307 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812337 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812349 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812191 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812265 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812334 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812184 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812354 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812208 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812230 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812377 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812181 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812314 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812350 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812229 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812225 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21812295 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21812353 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21812367 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21812340 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21812289 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02241752 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01672776 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21812380 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21812318 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02120883 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21812379 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21812332 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21812314 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01199375 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02120884 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21812303 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02142092 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21812285 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01426302 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21812357 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21812288 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02241750 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02142080 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01650613 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21812297 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01590805 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21812298 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21812308 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21812373 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01672773 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21812295 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21812345 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01672777 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01509857 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21812361 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21812381 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02142089 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21812283 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21812287 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21812334 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21812349 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21812359 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21812310 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01840613 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21812339 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01968372 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21812302 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21812343 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21812322 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02120882 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21812354 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21812307 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21812356 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21812344 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21812280 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21812370 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21812282 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21812291 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21812316 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21812313 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01649757 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21812364 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21812278 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21812324 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10376143 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10489365 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10489367 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10489366 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1416856_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10376142 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10489364 | - | - |
| Mouse Genome 430 2.0 Array | 1416856_at | - | - |
| Mouse Genome 430 2.0 Array | 1416856_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200043408.mm.1 | NM_025699.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200043408.mm.1 | NM_025699.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200043408.mm.1 | TC0200005090.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1416856_PM_at | - | - |
| Mouse Expression Set 430 | 1416856_at | - | - |
| Murine Genome U74v2 Set | 160200_at | - | - |
| Murine Genome U74v2 Set | 160200_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416856_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416856_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416856_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416856_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 360079 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000360079 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2684413 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2684413 | - | - |
| mouse-6 v1.1 (Illumina) | scl066680.1_207-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 360079 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl066680.1_207-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 360079 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2684413 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2684413 | - | - |

Oser1
Genomic Map