| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1500005522.mm.1 |
NM_019712.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500005522.mm.1 |
NM_019712.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500005522.mm.1 |
TC1500000721.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500005524.mm.1 |
NM_019712.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500005524.mm.1 |
NM_019712.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500005524.mm.1 |
TC1500000721.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500005516.mm.1 |
NM_019712.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500005516.mm.1 |
NM_019712.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500005516.mm.1 |
TC1500000721.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500005523.mm.1 |
NM_019712.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500005523.mm.1 |
NM_019712.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500005523.mm.1 |
TC1500000721.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500005520.mm.1 |
NM_019712.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500005520.mm.1 |
NM_019712.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500005520.mm.1 |
TC1500000721.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500005519.mm.1 |
NM_019712.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500005519.mm.1 |
NM_019712.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500005519.mm.1 |
TC1500000721.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500005521.mm.1 |
NM_019712.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500005521.mm.1 |
NM_019712.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500005521.mm.1 |
TC1500000721.mm.1 |
MTA TC |