Assay Details
| Gene Symbol: Hsd17b10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016763.2 | NP_058043.3 | 2 | 157 | 132 | |
| GenBank mRNA | AF233685.1 | - | 2 | 157 | 132 | |
| AK013340.1 | - | 2 | 153 | 132 | ||
| AK077735.1 | - | 2 | 151 | 132 | ||
| AK088685.1 | - | 2 | 173 | 132 | ||
| BC027517.1 | - | 2 | 157 | 3376098 | 132 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Hsd17b10
Genomic Map