Assay Details
| Gene Symbol: Ccr6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190333.1 | NP_001177262.1 | 2 - 3 | 384 | 73 | |
| NM_001190334.1 | NP_001177263.1 | 3 - 4 | 256 | 73 | ||
| NM_001190335.1 | NP_001177264.1 | 2 - 3 | 165 | 73 | ||
| NM_009835.4 | NP_033965.1 | 5 - 6 | 586 | 73 | ||
| GenBank mRNA | AK089712.1 | - | 5 - 6 | 587 | 73 | |
| AK154413.1 | - | 4 - 5 | 959 | 73 | ||
| BC100756.1 | - | 3 - 4 | 252 | 40046797 | 73 | |
| BC100757.1 | - | 2 - 3 | 161 | 40046796 | 73 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK089712.1, AK154413.1, BC100756.1, BC100757.1, NM_001190333.1, NM_001190334.1, NM_001190335.1, NM_009835.4 transcripts.
Set Membership: |
|

Ccr6
Genomic Map