Assay Details
| Gene Symbol: Pirb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011095.2 | NP_035225.2 | 13 - 14 | 2245 | 89 | |
| GenBank mRNA | AK142375.1 | - | 13 - 14 | 2226 | 89 | |
| AK152232.1 | - | 12 - 13 | 2448 | 89 | ||
| AK153116.1 | - | 12 - 13 | 2448 | 89 | ||
| AK161081.1 | - | 2216 | 89 | |||
| AK170720.1 | - | 10 - 11 | 2873 | 89 | ||
| BC026937.1 | - | 2199 | 4488338 | 89 | ||
| U83172.1 | - | 13 - 14 | 2189 | 89 | ||
| U96689.1 | - | 13 - 14 | 2228 | 89 | ||
| U96690.1 | - | 2180 | 89 | |||
| U96691.1 | - | 2192 | 89 | |||
| U96692.1 | - | 13 - 14 | 2229 | 89 | ||
| U96693.1 | - | 13 - 14 | 2245 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2097279 | U83172.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2097279 | AK161081.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2097279 | AK142375.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2097279 | NM_011095.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 365306 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 937815 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 929748 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 100040 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P124622 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2053398 | AK161081.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2097279 | U83172.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2097279 | AK161081.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2097279 | AK142375.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2097279 | NM_011095.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P445728 | U96692.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P445728 | U96693.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P445728 | U96691.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P445728 | U96689.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P445728 | BC026937.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P445726 | U96692.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P445726 | U96693.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P445726 | U96691.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P445726 | U96689.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P445726 | BC026937.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24947108 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24947114 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24947109 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24947101 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24947107 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24947104 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24947111 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24947112 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06112707 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P124622 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06112726 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06112746 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03806327 | - | - |
| Whole Mouse Genome G4122A | A_51_P445728 | U96692.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P445728 | U96693.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P445728 | U96691.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P445728 | U96689.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P445728 | BC026937.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P445726 | U96692.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P445726 | U96693.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P445726 | U96691.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P445726 | U96689.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P445726 | BC026937.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06112707 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P124622 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P03806327 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06112726 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06112746 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24946983 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24946920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24946991 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24946972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24946958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24946965 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24946969 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24946954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24946948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24946956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24946988 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24946913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24946911 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24946923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24946981 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24946974 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24946916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24946977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24946946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24946925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24946918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24946980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24946935 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24946967 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24946936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24946953 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24946945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24946952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24946979 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24946928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24946951 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24946931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24946926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24946949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24946985 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24946960 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24946985 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24946977 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24946960 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24946967 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P32979800 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24946979 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24946980 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24946973 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24946969 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24946974 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24946972 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24946963 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24946992 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24946983 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24946981 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24946986 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24946962 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24946991 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24946975 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24948379 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24946988 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10559447 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559450 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559451 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559448 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559477 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559452 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559459 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559469 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559461 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559462 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559470 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559453 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559474 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559456 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559472 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559449 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559458 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559465 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559475 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559464 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1424302_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10559446 | - | - |
| Mouse Genome 430 2.0 Array | 1424302_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1424302_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 18733_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 18733_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700019297.mm.1 | NM_011095.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700019297.mm.1 | NM_011095.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700019297.mm.1 | TC0700004631.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700019298.mm.1 | NM_011095.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700019298.mm.1 | NM_011095.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700019298.mm.1 | TC0700004631.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1424302_at | - | - |
| Mouse Expression Set 430 | 1424302_PM_at | - | - |
| Mouse Expression Set 430 | 1420464_PM_s_at | - | - |
| Mouse Expression Set 430 | 1420464_s_at | - | - |
| Murine Genome U74v2 Set | 100328_s_at | - | - |
| Murine Genome U74v2 Set | 98003_PM_at | - | - |
| Murine Genome U74v2 Set | 98003_at | - | - |
| Murine Genome U74v2 Set | 100328_PM_s_at | - | - |
| Murine Genome U74v2 Set | 168294_PM_f_at | - | - |
| Murine Genome U74v2 Set | 168294_f_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424302_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424302_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420464_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420464_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424302_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424302_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6130022 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6130022 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006130022 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1260379 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1260379 | - | - |
| mouse-6 v1.1 (Illumina) | scl018733.1_24-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6130022 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl018733.1_24-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6130022 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1260379 | - | - |

Pirb
Genomic Map