Assay Details
| Gene Symbol: Ube2j2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021402.6 | NP_067377.4 | 1 - 2 | 340 | 150 | |
| GenBank mRNA | AK076265.1 | - | 1 - 2 | 304 | 151 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK076265.1, NM_021402.6 transcripts.
Set Membership: |
|

Ube2j2
Genomic Map