Assay Details
| Gene Symbol: Paqr8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028829.3 | NP_083105.3 | 1 - 2 | 64 | 81 | |
| GenBank mRNA | AK029772.1 | - | 1 - 2 | 81 | 81 | |
| AK031969.1 | - | 1 - 2 | 77 | 81 | ||
| AK048045.1 | - | 1 - 2 | 79 | 81 | ||
| AK147376.1 | - | 1 - 2 | 69 | 81 | ||
| AY424297.1 | - | 1 - 2 | 79 | 81 | ||
| BC059813.1 | - | 1 - 2 | 64 | 6843180 | 81 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK031969.1, AK048045.1, AK147376.1, AY424297.1, BC059813.1, NM_028829.3 transcripts.
Set Membership: |
|

Paqr8
Genomic Map