Assay Details
| Gene Symbol: Tktl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028927.3 | NP_083203.2 | 1 | 1949 | 82 | |
| GenBank mRNA | AK016603.1 | - | 1 | 82 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Tktl2
Genomic Map