Assay Details
| Gene Symbol: Mettl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK043389.1 | - | 1 - 2 | 440 | 77 | |
| AK080683.1 | - | 1 - 2 | 511 | 77 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK043389.1, AK080683.1 transcripts.
Set Membership: |
|

Mettl4
Genomic Map