Assay Details
| Gene Symbol: Prss35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178738.3 | NP_848853.2 | 1 - 2 | 181 | 96 | |
| GenBank mRNA | AK030671.1 | - | 1 - 2 | 166 | 96 | |
| AK031411.1 | - | 1 - 2 | 166 | 96 | ||
| AK031644.1 | - | 1 - 2 | 166 | 96 | ||
| AK076281.1 | - | 1 - 2 | 166 | 96 | ||
| BC075675.1 | - | 1 - 2 | 150 | 30547401 | 96 | |
| DQ223037.1 | - | 1 - 2 | 141 | 96 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK030671.1, AK031411.1, AK031644.1, BC075675.1, DQ223037.1, NM_178738.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2111985 | DQ223037.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2111985 | BC075675.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2111985 | AK031644.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2111985 | AK076281.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2111985 | AK030671.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 931736 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 193043 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 274506 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 951494 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 143622 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 580350 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 206637 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 325011 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 57295 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 2293 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 84390 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P125988 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P157772 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2111985 | DQ223037.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2111985 | BC075675.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2111985 | AK031644.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2111985 | AK076281.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2111985 | AK030671.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P420600 | DQ223037.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P420600 | BC075675.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P420600 | AK031644.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P420600 | AK031411.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26700952 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26700940 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26700939 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26700933 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26700946 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26700937 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26700947 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26700941 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26700934 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26700938 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26700945 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26700951 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26700953 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26700949 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26700950 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P157772 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P125988 | - | - |
| Whole Mouse Genome G4122A | A_51_P420600 | DQ223037.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P420600 | BC075675.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P420600 | AK031644.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P420600 | AK031411.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P121940 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P109981 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P157772 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P125988 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07515119 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07515115 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07515110 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07515116 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26700946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26700955 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26700969 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26701003 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26701048 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26700965 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26700945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26700937 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26701010 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26700957 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26700947 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26700977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26701029 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26701018 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26700938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26701009 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26701036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26701014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26701006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26701022 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26701046 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26701000 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26701050 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26700934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26701011 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26700997 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26700983 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26701008 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26701038 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26701020 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26700939 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26701028 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26700967 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26701043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26701041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26700992 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26700961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26700971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26700988 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26700950 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26700984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26700980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26700970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26700949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26700942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26700933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26700952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26700958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26701049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26700999 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26700995 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26700941 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26700990 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26700974 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26700993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26701024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26700968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26701027 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26700963 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26700960 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26700974 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07471441 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26700971 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07849338 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26700967 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07515107 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26700932 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26700937 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26700957 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08145759 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26701033 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26701036 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26700977 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26700983 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07515118 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26700949 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26700934 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26700947 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08113599 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26700933 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26700958 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26700950 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26700953 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26701029 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07463513 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26700969 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26701041 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26701038 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07515112 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26700980 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26700945 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07849348 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26700946 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26700955 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08053960 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26700965 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26700961 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26701028 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26700962 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10587618 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10587619 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10587617 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10587620 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1434195_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10587616 | - | - |
| Mouse Genome 430 2.0 Array | 1434195_at | - | - |
| Mouse Genome 430 2.0 Array | 1434195_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 244954_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 244954_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0900008794.mm.1 | TC0900001119.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900008796.mm.1 | TC0900001119.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900008795.mm.1 | TC0900001119.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900008797.mm.1 | TC0900001119.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900008791.mm.1 | NM_178738.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900008791.mm.1 | NM_178738.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900008791.mm.1 | TC0900001119.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900008792.mm.1 | NM_178738.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900008792.mm.1 | NM_178738.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900008792.mm.1 | TC0900001119.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1434195_at | - | - |
| Mouse Expression Set 430 | 1434195_PM_at | - | - |
| Murine Genome U74v2 Set | 165432_i_at | - | - |
| Murine Genome U74v2 Set | 165432_PM_i_at | - | - |
| Mouse Genome 430A 2.0 Array | 1434195_at | - | - |
| Mouse Genome 430A 2.0 Array | 1434195_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1434195_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1434195_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 396058 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 100050114 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 100050114 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3450725 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003450725 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2609897 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2609897 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1226541 | - | - |
| mouse-6 v1.1 (Illumina) | scl36644.3_0-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 100050114 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3450725 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl36644.3_0-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3450725 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 100050114 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2609897 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1226541 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2609897 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl36644.3_0-S | - | - |

Prss35
Genomic Map