Assay Details
| Gene Symbol: Mgat4e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145300.2 | NP_001138772.1 | 2 - 3 | 510 | 81 | |
| GenBank mRNA | AK016512.1 | - | 2 - 3 | 512 | 80 | |
| BY716191.1 | - | 2 - 3 | 512 | 80 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK016512.1 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
carbohydrate metabolic process
biological_process
alpha-1,6-mannosyltransferase activity
alpha-1,2-mannosyltransferase activity
mannosyltransferase activity
alpha-1,3-mannosyltransferase activity
alpha-1,3-galactosyltransferase activity
molecular_function
UDP-glucose:glycoprotein glucosyltransferase activity
glycolipid mannosyltransferase activity
oligosaccharyl transferase activity
dolichyl-phosphate-glucose-glycolipid alpha-glucosyltransferase activity
acetylglucosaminyltransferase activity
acetylgalactosaminyltransferase activity
galactosyltransferase activity
fucosyltransferase activity
O antigen polymerase activity
lipopolysaccharide-1,6-galactosyltransferase activity
transferase activity
transferase activity, transferring glycosyl groups
transferase activity, transferring hexosyl groups
cellulose synthase activity
9-phenanthrol UDP-glucuronosyltransferase activity
1-phenanthrol glycosyltransferase activity
9-phenanthrol glycosyltransferase activity
1,2-dihydroxy-phenanthrene glycosyltransferase activity
phenanthrol glycosyltransferase activity
beta-1,4-mannosyltransferase activity
alpha-1,2-galactosyltransferase activity
dolichyl pyrophosphate Man7GlcNAc2 alpha-1,3-glucosyltransferase activity
lipopolysaccharide-1,5-galactosyltransferase activity
dolichyl pyrophosphate Man9GlcNAc2 alpha-1,3-glucosyltransferase activity
dolichyl pyrophosphate Glc1Man9GlcNAc2 alpha-1,3-glucosyltransferase activity
inositol phosphoceramide synthase activity
glucosyltransferase activity
alpha-(1->3)-fucosyltransferase activity
alpha-(1->6)-fucosyltransferase activity
indole-3-butyrate beta-glucosyltransferase activity
salicylic acid glucosyltransferase (ester-forming) activity
salicylic acid glucosyltransferase (glucoside-forming) activity
benzoic acid glucosyltransferase activity
chondroitin hydrolase activity
dolichyl-pyrophosphate Man7GlcNAc2 alpha-1,6-mannosyltransferase activity
cytokinin 9-beta-glucosyltransferase activity
carbohydrate metabolic process
biological_process
alpha-1,6-mannosyltransferase activity
alpha-1,2-mannosyltransferase activity
mannosyltransferase activity
alpha-1,3-mannosyltransferase activity
alpha-1,3-galactosyltransferase activity
molecular_function
UDP-glucose:glycoprotein glucosyltransferase activity
glycolipid mannosyltransferase activity
oligosaccharyl transferase activity
dolichyl-phosphate-glucose-glycolipid alpha-glucosyltransferase activity
acetylglucosaminyltransferase activity
acetylgalactosaminyltransferase activity
galactosyltransferase activity
fucosyltransferase activity
O antigen polymerase activity
lipopolysaccharide-1,6-galactosyltransferase activity
transferase activity
transferase activity, transferring glycosyl groups
transferase activity, transferring hexosyl groups
cellulose synthase activity
9-phenanthrol UDP-glucuronosyltransferase activity
1-phenanthrol glycosyltransferase activity
9-phenanthrol glycosyltransferase activity
1,2-dihydroxy-phenanthrene glycosyltransferase activity
phenanthrol glycosyltransferase activity
beta-1,4-mannosyltransferase activity
alpha-1,2-galactosyltransferase activity
dolichyl pyrophosphate Man7GlcNAc2 alpha-1,3-glucosyltransferase activity
lipopolysaccharide-1,5-galactosyltransferase activity
dolichyl pyrophosphate Man9GlcNAc2 alpha-1,3-glucosyltransferase activity
dolichyl pyrophosphate Glc1Man9GlcNAc2 alpha-1,3-glucosyltransferase activity
inositol phosphoceramide synthase activity
glucosyltransferase activity
alpha-(1->3)-fucosyltransferase activity
alpha-(1->6)-fucosyltransferase activity
indole-3-butyrate beta-glucosyltransferase activity
salicylic acid glucosyltransferase (ester-forming) activity
salicylic acid glucosyltransferase (glucoside-forming) activity
benzoic acid glucosyltransferase activity
chondroitin hydrolase activity
dolichyl-pyrophosphate Man7GlcNAc2 alpha-1,6-mannosyltransferase activity
cytokinin 9-beta-glucosyltransferase activity
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P192924 | NR_026896.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P192924 | AK016512.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P192924 | BC145149.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P192924 | BX630257.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P192924 | NM_001145300.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P192924 | NR_026896.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P192924 | AK016512.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P192924 | BC145149.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P192924 | BX630257.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P192924 | NM_001145300.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 6121 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 65166 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 68121 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 288269 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 785534 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P192924 | NR_026896.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P192924 | AK016512.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P192924 | BC145149.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P192924 | BX630257.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P192924 | NM_001145300.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P192924 | NR_026896.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P192924 | AK016512.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P192924 | BC145149.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P192924 | BX630257.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P192924 | NM_001145300.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P192924 | NR_026896.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P192924 | AK016512.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P192924 | BC145149.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P192924 | BX630257.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P192924 | NM_001145300.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P192924 | NR_026896.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P192924 | AK016512.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P192924 | BC145149.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P192924 | BX630257.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P192924 | NM_001145300.1 | mouse_transcriptome |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20633953 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20633890 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20633955 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00393362 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20633921 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20633933 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20633920 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20633960 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01095412 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20633939 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20633930 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20633951 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00682545 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20633936 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20633938 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20633926 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00619837 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20633922 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20633949 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00118634 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20633892 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20633934 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20633925 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20633923 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00013316 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20633945 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00688046 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00753797 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20633950 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20633946 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00998093 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20633940 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20633962 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20633929 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10357929 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10357930 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10357928 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1430094_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10357927 | - | - |
| Mouse Genome 430 2.0 Array | 1430094_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1430094_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 71001_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 71001_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0100025075.mm.1 | NM_001145300.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100025075.mm.1 | TC0100003111.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1430094_at | - | - |
| Mouse Expression Set 430 | 1430094_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1430094_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1430094_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1213572 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1213572 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1213572 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1213572 | - | - |

Mgat4e
Genomic Map