Assay Details
| Gene Symbol: Zc3h4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198631.2 | NP_941033.2 | 1 - 2 | 116 | 82 | |
| GenBank mRNA | BC057627.1 | - | 1 - 2 | 61 | 6412890 | 82 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC057627.1, NM_198631.2 transcripts.
Set Membership: |
|

Zc3h4
Genomic Map