Assay Details
| Gene Symbol: Zc3h4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198631.2 | NP_941033.2 | 2 - 3 | 334 | 74 | |
| GenBank mRNA | AK138078.1 | - | 1 - 2 | 2152 | 74 | |
| AK151148.1 | - | 3 - 4 | 386 | 74 | ||
| AK151306.1 | - | 3 - 4 | 386 | 74 | ||
| BC057627.1 | - | 2 - 3 | 279 | 6412890 | 74 | |
| BY167917.1 | - | 3 - 4 | 383 | 74 | ||
| CJ101946.1 | - | 2 - 3 | 339 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zc3h4
Genomic Map