Assay Details
| Gene Symbol: Zc3h4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198631.2 | NP_941033.2 | 3 - 4 | 442 | 68 | |
| GenBank mRNA | AK151148.1 | - | 4 - 5 | 494 | 68 | |
| AK151306.1 | - | 4 - 5 | 494 | 68 | ||
| BC057627.1 | - | 3 - 4 | 387 | 6412890 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zc3h4
Genomic Map