| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1100034500.mm.1 |
NM_201255.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100034500.mm.1 |
NM_201255.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100034500.mm.1 |
TC1100003759.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100034499.mm.1 |
NM_201255.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100034499.mm.1 |
NM_201255.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100034499.mm.1 |
TC1100003759.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100034501.mm.1 |
NM_201255.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100034501.mm.1 |
NM_201255.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100034501.mm.1 |
TC1100003759.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100034502.mm.1 |
NM_201255.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100034502.mm.1 |
NM_201255.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100034502.mm.1 |
TC1100003759.mm.1 |
MTA TC |