Assay Details
| Gene Symbol: Kcnk18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207261.3 | NP_997144.1 | 1 - 2 | 338 | 66 | |
| GenBank mRNA | AK139600.1 | - | 1 - 2 | 772 | 66 | |
| AK162136.1 | - | 1 - 2 | 341 | 66 | ||
| AY325301.1 | - | 1 - 2 | 252 | 66 | ||
| AY542902.1 | - | 1 - 2 | 252 | 66 | ||
| BC104132.1 | - | 1 - 2 | 252 | 66 | ||
| BC104133.1 | - | 1 - 2 | 249 | 66 | ||
| BC115705.1 | - | 1 - 2 | 251 | 66 | ||
| BC115887.1 | - | 1 - 2 | 245 | 66 | ||
| BC127136.1 | - | 1 - 2 | 252 | 40045157 | 66 | |
| BC127137.1 | - | 1 - 2 | 252 | 40045158 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kcnk18
Genomic Map