| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0X00010508.mm.1 |
NM_009031.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00010508.mm.1 |
NM_009031.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00010508.mm.1 |
TC0X00001651.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00010496.mm.1 |
NM_009031.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00010496.mm.1 |
NM_009031.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00010496.mm.1 |
TC0X00001651.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00010502.mm.1 |
NM_009031.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00010502.mm.1 |
NM_009031.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00010502.mm.1 |
TC0X00001651.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00010505.mm.1 |
NM_009031.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00010505.mm.1 |
NM_009031.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00010505.mm.1 |
TC0X00001651.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00010506.mm.1 |
NM_009031.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00010506.mm.1 |
NM_009031.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00010506.mm.1 |
TC0X00001651.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00010504.mm.1 |
NM_009031.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00010504.mm.1 |
NM_009031.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00010504.mm.1 |
TC0X00001651.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00010500.mm.1 |
NM_009031.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00010500.mm.1 |
NM_009031.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00010500.mm.1 |
TC0X00001651.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00010503.mm.1 |
NM_009031.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00010503.mm.1 |
NM_009031.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00010503.mm.1 |
TC0X00001651.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00010509.mm.1 |
NM_009031.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00010509.mm.1 |
NM_009031.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00010509.mm.1 |
TC0X00001651.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00010511.mm.1 |
NM_009031.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00010511.mm.1 |
NM_009031.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00010511.mm.1 |
TC0X00001651.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00010507.mm.1 |
NM_009031.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00010507.mm.1 |
NM_009031.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00010507.mm.1 |
TC0X00001651.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00010510.mm.1 |
NM_009031.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00010510.mm.1 |
NM_009031.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00010510.mm.1 |
TC0X00001651.mm.1 |
MTA TC |