| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0X00010517.mm.1 |
NM_009031.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00010517.mm.1 |
NM_009031.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00010517.mm.1 |
TC0X00001651.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00010515.mm.1 |
NM_009031.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00010515.mm.1 |
NM_009031.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00010515.mm.1 |
TC0X00001651.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00010512.mm.1 |
NM_009031.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00010512.mm.1 |
NM_009031.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00010512.mm.1 |
TC0X00001651.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00010513.mm.1 |
NM_009031.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00010513.mm.1 |
NM_009031.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00010513.mm.1 |
TC0X00001651.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00010518.mm.1 |
NM_009031.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00010518.mm.1 |
NM_009031.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00010518.mm.1 |
TC0X00001651.mm.1 |
MTA TC |