Assay Details
| Gene Symbol: Gpank1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032460.2 | NP_115849.2 | 1 | 44 | 123 | |
| GenBank mRNA | AK009290.1 | - | 123 | |||
| BC120637.1 | - | 1 | 45 | 40129560 | 123 | |
| BC120639.1 | - | 1 | 45 | 40129562 | 123 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC120637.1, BC120639.1, NM_032460.2 transcripts.
Set Membership: |
|

Gpank1
Genomic Map