Assay Details
| Gene Symbol: BC051408 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC051408.1 | - | 1 - 2 | 167 | 88 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 33228 |
Gm33228 | -2.5mb | Sense | Chr. 18 - 25821269 - 26216558 on NCBI Build GRCm38 | |
| Downstream | RIKEN cDNA 4930578E11 gene | 4930578E11Rik | 670.7kb | Anti-sense | Chr. 18 - 29396716 - 29496416 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

BC051408
Genomic Map