Assay Details
| Gene Symbol: 4930448C13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045960.1 | 2 - 3 | 121 | |||
| GenBank mRNA | AK015413.1 | - | 2 - 3 | 121 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 16497 | Gm16497 | ENSMUSG00000066623 | -736.7kb | Anti-sense | Chr. 12 - 14261534 - 14285684 on NCBI Build GRCm38 |
| Downstream | tribbles homolog 2 (Drosophila) | Trib2 |
AW319517, TRB-2, TRB2 |
742.3kb | Sense | Chr. 12 - 15791725 - 15816962 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

4930448C13Rik
Genomic Map