Assay Details
| Gene Symbol: 4930448C13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045960.1 | 6 - 7 | 478 | 92 | ||
| GenBank mRNA | AK015413.1 | - | 6 - 7 | 480 | 92 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 16497 | Gm16497 | ENSMUSG00000066623 | -712.2kb | Anti-sense | Chr. 12 - 14261534 - 14285684 on NCBI Build GRCm38 |
| Downstream | tribbles homolog 2 (Drosophila) | Trib2 |
AW319517, TRB-2, TRB2 |
782.5kb | Sense | Chr. 12 - 15791725 - 15816962 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 140793 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 476786 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 148041 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 27367 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 837633 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 759212 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 101728 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 212092 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 192899 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 338034 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 704312 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 402942 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 966388 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 890637 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 827519 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 83683 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 108060 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07546828 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07546889 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07546866 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P07546828 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P07546866 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28257067 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28257058 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28256991 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28256983 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28257104 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28257103 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28256912 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28256908 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28257060 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28256904 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28257101 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28257051 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28257053 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28256943 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28256950 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28256954 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28257056 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P10393092 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28256946 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28257057 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28257082 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P10372698 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28256896 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28256982 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28257092 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28257098 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28256985 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28257050 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09976096 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28257080 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28256910 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28256980 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28257105 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28256994 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28257046 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28256986 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28256906 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28257083 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P10255207 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28257065 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28256899 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28257076 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28257073 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28257063 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28257055 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28257070 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28257099 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28257102 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P10205704 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28256944 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28257071 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28256911 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28256955 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28256901 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09998031 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28257068 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28256939 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28256949 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28256898 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28257061 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28256894 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28257087 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28256988 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28257078 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28256951 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28256905 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28257052 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Clariom S Array | PSR1200010121.mm.1 | NR_045960.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200010121.mm.1 | NR_045960.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200010121.mm.1 | TC1200001447.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200010122.mm.1 | NR_045960.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200010122.mm.1 | NR_045960.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200010122.mm.1 | TC1200001447.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 863480 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 102650722 | - | - |

4930448C13Rik
Genomic Map