Assay Details
| Gene Symbol: Gm32828 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK043922.1 | - | 2 - 3 | 241 | 66 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 1700101O22 gene | 1700101O22Rik | -375.1kb | Sense | Chr. 12 - 7372039 - 7380330 on NCBI Build GRCm38 | |
| Downstream | apolipoprotein B | Apob |
AI315052, Apo B-100, apob-100, apob-48 |
221.7kb | Anti-sense | Chr. 12 - 7977633 - 8016839 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gm32828
Genomic Map