Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | homeobox A9 | Hoxa9 |
D6a9, Hox-1.7 |
-877.0 | Sense | Chr. 6 - 52223097 - 52227370 on NCBI Build GRCm38 |
| Downstream | microRNA 196b | Mir196b |
Mirn196b, mir-196b |
1.2kb | Sense | Chr. 6 - 52230081 - 52230165 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map