Assay Details
| Gene Symbol: 1500035N22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK005363.1 | - | 2 | 433 | 122 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
protein kinase, AMP-activated, gamma 2 non-catalytic subunit |
Prkag2 |
2410051C13Rik, AAKG, AAKG2, AI854673, H91620p, WPWS |
Anti-sense | Chr. 5 - 24862735 - 25101906 on NCBI Build GRCm38 | |
| Downstream | RIKEN cDNA 2900005J15 gene | 2900005J15Rik | 102.8kb | Sense | Chr. 5 - 25100975 - 25103007 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

1500035N22Rik
Genomic Map