Assay Details
| Gene Symbol: LOC545403 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK082173.1 | - | 1 - 2 | 62 | 84 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC102632465 | LOC102632465 | -405.7kb | Sense | Chr. 2 - 6466079 - 6478619 on NCBI Build GRCm38 | |
| Downstream | RIKEN cDNA 5031426D15 gene | 5031426D15Rik | 35.8kb | Anti-sense | Chr. 2 - 6922702 - 6928722 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC545403
Genomic Map