Assay Details
| Gene Symbol: AI314760 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK040485.1 | - | 1 - 2 | 1563 | 83 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
casein kinase 1, alpha 1 |
Csnk1a1 |
2610208K14Rik, 4632404G05Rik, 5430427P18Rik, CK1a, Csnk1a |
Sense | Chr. 18 - 61555165 - 61590315 on NCBI Build GRCm38 | |
| Downstream | braveheart long non-coding RNA | Bvht |
Braveheart, Gm20748 |
54.1kb | Sense | Chr. 18 - 61639653 - 61647485 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

AI314760
Genomic Map