Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 9130209A04 gene | 9130209A04Rik | -6.6kb | Sense | Chr. 18 - 47452668 - 47470868 on NCBI Build GRCm38 | |
| Downstream | predicted gene 5095 | Gm5095 | A730092B10 | 44.5kb | Sense | Chr. 18 - 47537735 - 47718636 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Genomic Map