Assay Details
| Gene Symbol: 1700101I19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK007108.1 | - | 1 | 247 | 134 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
protease, serine 40 |
Prss40 | Tesp2 | -18.2kb | Sense | Chr. 1 - 34536515 - 34560993 on NCBI Build GRCm38 |
| Downstream | PHD finger protein 19 pseudogene | AA619741 | 53.9kb | Sense | Chr. 1 - 34633535 - 34636210 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

1700101I19Rik
Genomic Map