Assay Details
| Gene Symbol: Cramp1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020608.3 | NP_065633.2 | 14 - 15 | 2908 | 88 | |
| GenBank mRNA | AK089393.1 | - | 14 - 15 | 3168 | 88 | |
| AK129356.1 | - | 5 - 6 | 1084 | 88 | ||
| AK147411.1 | - | 14 - 15 | 2909 | 88 | ||
| AK147449.1 | - | 14 - 15 | 2909 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cramp1l
Genomic Map