Assay Details
| Gene Symbol: Gm38569 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK042846.1 | - | 1 - 2 | 497 | 89 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 4732471J01 gene | 4732471J01Rik | -17.8kb | Sense | Chr. 7 - 25376819 - 25566417 on NCBI Build GRCm38 | |
| Downstream | glutamate rich 4 | Erich4 |
EG632778, Gm7092, Kseg1 |
19.8kb | Anti-sense | Chr. 7 - 25614621 - 25615892 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gm38569
Genomic Map