Assay Details
| Gene Symbol: Map3k10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290528.1 | NP_001277457.1 | 1 - 2 | 1236 | 66 | |
| GenBank mRNA | AK053830.1 | - | 1 - 2 | 1236 | 66 | |
| BC078445.1 | - | 1 - 2 | 275 | 66 | ||
More Information
Set Membership: |
|

Map3k10
Genomic Map