Assay Details
| Gene Symbol: Gsk3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031667.1 | NP_001026837.1 | 3 - 4 | 746 | 97 | |
| GenBank mRNA | BQ885209.1 | - | 3 - 4 | 339 | 97 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gsk3a
Genomic Map