Assay Details
| Gene Symbol: 1700025K04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK005576.1 | - | 1 - 2 | 59 | 75 | |
| AK006351.1 | - | 85 | 75 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 1700021G15 gene | 1700021G15Rik | -114.8kb | Sense | Chr. 10 - 126255117 - 126257221 on NCBI Build GRCm38 | |
| Downstream | microRNA 378d | Mir378d |
Gm27650, mmu-mir-378d |
336.0kb | Anti-sense | Chr. 10 - 126710282 - 126710391 on NCBI Build GRCm38 |
More Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2370210 | AK006351.1 | mouse_transcriptome |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27530218 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27530239 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27530257 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27530199 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08675533 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27530246 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08564377 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08344396 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27530217 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27530185 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27530208 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27530253 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27530197 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27530193 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27530232 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27530202 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27530196 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27530180 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27530249 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08853358 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27530214 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27530251 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27530147 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27530206 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27530255 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08853349 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27530216 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27530210 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27530153 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27530203 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27530190 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27530265 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08853352 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27530192 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27530262 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27530261 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27530204 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27530178 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27530173 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27530223 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27530188 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27530273 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08872836 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27530159 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27530150 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08473310 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27530242 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27530212 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27530233 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27530229 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27530186 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27530268 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27530225 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27530264 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27530201 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27530259 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27530158 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08352155 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27530222 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27530175 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27530149 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27530272 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27530162 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27530169 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27530241 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27530179 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27530152 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27530156 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27530235 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27530238 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27530171 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27530184 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27530254 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT MG-430 PM Array Plate | 1454165_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1454165_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1454165_at | - | - |
| Mouse Expression Set 430 | 1454165_PM_at | - | - |
| Mouse Expression Set 430 | 1454165_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1454165_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1454165_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 852083 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 106400731 | - | - |

1700025K04Rik
Genomic Map