Assay Details
| Gene Symbol: 2700012I20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK012238.1 | - | 1 - 2 | 61 | 95 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 10658 | Gm10658 | ENSMUSG00000074284 | -3.3kb | Sense | Chr. 9 - 57056897 - 57071966 on NCBI Build GRCm38 |
| Downstream |
mannosidase, alpha, class 2C, member 1, opposite strand |
Man2c1os |
2410133F24Rik, AI851351 |
54.0kb | Sense | Chr. 9 - 57129982 - 57131290 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

2700012I20Rik
Genomic Map