Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | paired box 6 | Pax6 |
1500038E17Rik, AEY11, Dey, Gsfaey11, Pax-6, Sey |
Anti-sense | Chr. 2 - 105667518 - 105697364 on NCBI Build GRCm38 | |
| Downstream | elongation protein 4 homolog (S. cerevisiae) | Elp4 |
A330107A17Rik, Paxneb |
25.7kb | Sense | Chr. 2 - 105700109 - 105904500 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Genomic Map