Assay Details
| Gene Symbol: Gm38572 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK054526.1 | - | 2 - 3 | 202 | 68 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 7420700N18 gene | 7420700N18Rik | -241.9kb | Anti-sense | Chr. 8 - 31367749 - 31382439 on NCBI Build GRCm38 | |
| Downstream | neuregulin 1 | Nrg1 |
6030402G23Rik, ARIA, D230005F13Rik, GGF, GGFII, HRG, HRGalpha, Hgl, NDF, SMDF |
148.0kb | Sense | Chr. 8 - 31807452 - 32890583 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm38572
Genomic Map