Assay Details
| Gene Symbol: Gm5441 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK039060.1 | - | 1 - 2 | 147 | 90 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
protein tyrosine phosphatase, receptor type, N polypeptide 2 |
Ptprn2 |
4930425H11Rik, IA2beta, PTP-NP, Phol, mKIAA0387, phogrin |
-4.4kb | Anti-sense | Chr. 12 - 116485692 - 117278167 on NCBI Build GRCm38 |
| Downstream |
predicted gene, 39559 |
Gm39559 | 8.5kb | Anti-sense | Chr. 12 - 117345562 - 117347995 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm5441
Genomic Map