Assay Details
| Gene Symbol: Gm5142 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004158.2 | NP_001004158.1 | 1 - 2 | 419 | 89 | |
| GenBank mRNA | AK076969.1 | - | 1 - 2 | 420 | 89 | |
| BC099536.1 | - | 1 - 2 | 391 | 30918677 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P1036735 | NM_001004158.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P1036735 | AK076969.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P1036735 | BC099536.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P1036735 | NM_001004158.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P1036735 | AK076969.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P1036735 | BC099536.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 153390 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 864507 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 259053 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 223168 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 788453 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 143640 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 877003 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 149228 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 157384 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P175438 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P160348 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P1036735 | NM_001004158.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P1036735 | AK076969.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P1036735 | BC099536.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P1036735 | NM_001004158.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P1036735 | AK076969.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P1036735 | BC099536.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29637282 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29637279 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29637273 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29637289 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29637286 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29637287 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29637274 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29637276 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29637284 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29637278 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29637281 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P160348 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P175438 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08127320 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02859585 | - | - |
| Whole Mouse Genome G4122A | A_52_P1036735 | NM_001004158.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P1036735 | AK076969.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P1036735 | BC099536.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P1036735 | NM_001004158.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P1036735 | AK076969.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P1036735 | BC099536.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P175438 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P160348 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11589602 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11589606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29637284 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29637287 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29637236 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29637239 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29637282 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29637232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29637278 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29637249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29637238 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29637276 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29637294 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29637270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29637244 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29637227 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29637234 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29637246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29637241 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29637254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29637243 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29637296 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29637224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29637250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29637281 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29637258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29637274 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29637280 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29637273 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29637237 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29637279 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29637259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29637286 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29637290 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11589612 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29637282 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29637214 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29637279 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29637291 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29637280 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29637286 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11541718 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29637284 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29637287 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29637213 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11589606 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29637278 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29637276 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29637274 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10420482 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10420481 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10420480 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1445441_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10420479 | - | - |
| Mouse Genome 430 2.0 Array | 1445441_at | - | - |
| Mouse Genome 430 2.0 Array | 1445441_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 380907_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 380907_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1400016656.mm.1 | NM_001004158.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400016656.mm.1 | NM_001004158.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400016656.mm.1 | TC1400002159.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1400016654.mm.1 | NM_001004158.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400016654.mm.1 | NM_001004158.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400016654.mm.1 | TC1400002159.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1400016655.mm.1 | NM_001004158.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400016655.mm.1 | NM_001004158.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400016655.mm.1 | TC1400002159.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1445441_PM_at | - | - |
| Mouse Expression Set 430 | 1445441_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1445441_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1445441_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 336968 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1212987 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2960992 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2960992 | - | - |

Gm5142
Genomic Map