Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | methionine sulfoxide reductase A | Msra |
2310045J23Rik, 6530413P12Rik, MSR-A |
-76.2kb | Sense | Chr. 14 - 64122621 - 64455903 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA A930011O12 gene | A930011O12Rik |
Rncr3, mir-124-1, mir-3078 |
55.3kb | Anti-sense | Chr. 14 - 64588115 - 64593961 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Genomic Map