Assay Details
| Gene Symbol: 4930426I24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK015206.1 | - | 1 - 2 | 497 | 75 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc finger and BTB domain containing 1 | Zbtb1 |
9430077A12, AI256369, Bsg2, C430003J21Rik |
-5.8kb | Anti-sense | Chr. 12 - 76370192 - 76396955 on NCBI Build GRCm38 |
| Downstream | heat shock protein 2 | Hspa2 |
70kDa, HSP70.2, HSP70A2, Hsp70-2 |
266.0 | Anti-sense | Chr. 12 - 76403844 - 76406938 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

4930426I24Rik
Genomic Map